siRNA Details
| Gene Symbol: MS4A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021201.4 | NP_067024.1 | 7 | 2366 | ||
| NM_206938.1 | NP_996821.1 | 6 | 2231 | |||
| NM_206939.1 | NP_996822.1 | 7 | 2318 | |||
| NM_206940.1 | NP_996823.1 | 6 | 2183 | |||

MS4A7
Genomic Map