siRNA Details
| Gene Symbol: TP53INP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005260476.2 | XP_005260533.1 | 4 | 640 | ||
| XM_006723841.2 | XP_006723904.1 | 2 | 286 | |||
| XM_011528952.2 | XP_011527254.1 | 4 | 701 | |||
| XM_011528953.2 | XP_011527255.1 | 3 | 581 | |||
| GenBank mRNA | AJ297792.1 | - | 4 | 648 | ||
| AK292443.1 | - | 4 | 648 | |||
| AY422170.1 | - | 4 | 657 | |||
| BC035639.1 | - | 4 | 546 | 5547220 | ||

TP53INP2
Genomic Map