siRNA Details
| Gene Symbol: OPRPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302807.1 | NP_001289736.1 | 4 | 524 | ||
| NM_021225.4 | NP_067048.4 | 3 | 468 | |||
| NR_126503.1 | 2 | 402 | ||||
| GenBank mRNA | CD722615.1 | - | 3 | 468 | ||
| CD723077.1 | - | 4 | 451 | |||
| CK429697.1 | - | 2 | 319 | |||
| S83198.1 | - | 3 | 374 | |||

OPRPN
Genomic Map