siRNA Details
| Gene Symbol: USP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286457.1 | NP_001273386.1 | 6 | 710 | ||
| NM_001286458.1 | NP_001273387.1 | 7 | 885 | |||
| NM_001321858.1 | NP_001308787.1 | 6 | 636 | |||
| NM_003470.2 | NP_003461.2 | 6 | 841 | |||
| NR_135826.1 | 6 | 664 | ||||
| XM_017023652.1 | XP_016879141.1 | 6 | 1269 | |||
| XM_017023653.1 | XP_016879142.1 | 6 | 705 | |||
| GenBank mRNA | AK302771.1 | - | 6 | 642 | ||
| AK302872.1 | - | 6 | 664 | |||
| AK302912.1 | - | 7 | 829 | |||
| AK316441.1 | - | 6 | 711 | |||
| AY376241.1 | - | 6 | 672 | |||
| Z72499.1 | - | 6 | 841 | |||

USP7
Genomic Map