siRNA Details
| Gene Symbol: HRH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143828.1 | NP_001137300.1 | 2 | 1453 | ||
| NM_001160166.1 | NP_001153638.1 | 2 | 1553 | |||
| NM_021624.3 | NP_067637.2 | 3 | 1717 | |||
| GenBank mRNA | AF312230.1 | - | 3 | 1717 | ||
| BC112348.1 | - | 3 | 1677 | 40006540 | ||
More Information
Set Membership: |
|

HRH4
Genomic Map