siRNA Details
| Gene Symbol: GNB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021629.3 | NP_067642.1 | 10 | 1619 | ||
| XM_005247692.2 | XP_005247749.1 | 10 | 1540 | |||
| XM_006713721.2 | XP_006713784.1 | 10 | 1550 | |||
| GenBank mRNA | AK001890.1 | - | 10 | 1619 | ||
| AK022599.1 | - | 1 | 312 | |||
| AK297174.1 | - | 10 | 1416 | |||
| AK310009.1 | - | 5 | 889 | |||

GNB4
Genomic Map