siRNA Details
| Gene Symbol: C12orf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021640.3 | NP_067653.3 | 4 | 652 | ||
| GenBank mRNA | AF111805.1 | - | 2 | 244 | ||
| AF289485.1 | - | 4 | 620 | |||
| BC028904.1 | - | 3 | 556 | 4454943 | ||
| BC051871.1 | - | 4 | 611 | 5170816 | ||

C12orf10
Genomic Map