siRNA Details
| Gene Symbol: MFAP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297709.1 | NP_001284638.1 | 7 | 520 | ||
| NM_001297710.1 | NP_001284639.1 | 6 | 484 | |||
| NM_001297711.1 | NP_001284640.1 | 6 | 475 | |||
| NM_003480.3 | NP_003471.1 | 8 | 550 | |||
| NR_123733.1 | 8 | 550 | ||||
| NR_123734.1 | 7 | 520 | ||||
| GenBank mRNA | AK124368.1 | - | 6 | 471 | ||
| AK290585.1 | - | 8 | 341 | |||
| AK299475.1 | - | 7 | 348 | |||
| AK315807.1 | - | 8 | 337 | |||
| AU280458.1 | - | 8 | 488 | |||
| AY339060.1 | - | 6 | 256 | |||
| BC005901.1 | - | 8 | 340 | 4247343 | ||
| BU929788.1 | - | 7 | 324 | |||
| BX396041.2 | - | 6 | 283 | |||
| DN996275.1 | - | 8 | 372 | |||
| U37283.1 | - | 8 | 472 | |||

MFAP5
Genomic Map