siRNA Details
| Gene Symbol: NXF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022052.1 | NP_071335.1 | 18 | 1552 | ||
| GenBank mRNA | AF346619.1 | - | 18 | 1552 | ||
| AJ277527.1 | - | 18 | 1552 | |||
| AK057086.1 | - | 7 | 1935 | |||
| AK294826.1 | - | 579 | ||||
| BC031616.1 | - | 18 | 1545 | 5170537 | ||

NXF3
Genomic Map