siRNA Details
| Gene Symbol: ZNF335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022095.3 | NP_071378.1 | 8 | 1242 | ||
| XM_005260504.4 | XP_005260561.1 | 8 | 1955 | |||
| XM_011528979.2 | XP_011527281.1 | 7 | 1610 | |||
| XM_017028012.1 | XP_016883501.1 | 7 | 1013 | |||
| GenBank mRNA | AF395833.1 | - | 8 | 1219 | ||
| AK027358.1 | - | 1 | 543 | |||
| AK296989.1 | - | 5 | 813 | |||

ZNF335
Genomic Map