siRNA Details
| Gene Symbol: C11orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300913.1 | NP_001287842.1 | 5 | 1069 | ||
| NM_022338.3 | NP_071733.1 | 4 | 2074 | |||
| XM_005274053.3 | XP_005274110.1 | 4 | 2249 | |||
| XM_011545103.2 | XP_011543405.1 | 2 | 1669 | |||
| GenBank mRNA | AF264781.1 | - | 4 | 2034 | ||
| AF370372.1 | - | 3 | 1942 | |||
| AY358754.1 | - | 2 | 1636 | |||
| BC011765.2 | - | 3 | 1923 | 3614861 | ||

C11orf24
Genomic Map