siRNA Details
Gene Symbol: GPR87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023915.3 | NP_076404.3 | 3 | 685 | ||
GenBank mRNA | AB031548.1 | - | 3 | 617 | ||
AF237763.1 | - | 3 | 573 | |||
AF345566.1 | - | 1 | 363 | |||
AF411114.1 | - | 2 | 324 | |||
AK027784.1 | - | 3 | 685 | |||
BC009540.1 | - | 1 | 131 | |||
BC112941.1 | - | 3 | 594 | 4500655 | ||
BC117565.1 | - | 3 | 594 | 4521950 | ||
BC132888.1 | - | 3 | 651 | 40146910 | ||
BC132890.1 | - | 3 | 651 | 40146912 | ||
EU883576.1 | - | 2 | 324 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
