siRNA Details
| Gene Symbol: WDR18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024100.3 | NP_077005.2 | 10 | 1267 | ||
| XM_017027027.1 | XP_016882516.1 | 12 | 1793 | |||
| GenBank mRNA | AK131009.1 | - | 4 | 410 | ||
| BC000040.2 | - | 10 | 1252 | 3507673 | ||
| BC001648.2 | - | 10 | 1247 | 2819452 | ||

WDR18
Genomic Map