siRNA Details
| Gene Symbol: ATG16L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190266.1 | NP_001177195.1 | 11 | 1340 | ||
| NM_001190267.1 | NP_001177196.1 | 11 | 1338 | |||
| NM_017974.3 | NP_060444.3 | 10 | 1285 | |||
| NM_030803.6 | NP_110430.5 | 11 | 1342 | |||
| NM_198890.2 | NP_942593.2 | 7 | 853 | |||
| XM_005246082.2 | XP_005246139.1 | 12 | 1330 | |||
| XM_005246084.2 | XP_005246141.1 | 9 | 869 | |||
| XM_005246086.2 | XP_005246143.1 | 8 | 818 | |||
| XM_006712608.2 | XP_006712671.1 | 11 | 1141 | |||
| GenBank mRNA | AK000897.1 | - | 10 | 953 | ||
| AK001690.1 | - | 10 | 1130 | |||
| AK024453.1 | - | 10 | 1880 | |||
| AK027854.1 | - | 9 | 867 | |||
| AK308400.1 | - | 10 | 1183 | |||
| AY358182.1 | - | 11 | 1231 | |||
| AY398617.1 | - | 11 | 1231 | |||
| BC071846.1 | - | 7 | 691 | |||
| BC117337.1 | - | 11 | 1004 | |||
| BC143900.1 | - | 10 | 947 | |||
| EF079889.1 | - | 10 | 1028 | |||
| EF079890.1 | - | 7 | 596 | |||
| EF560734.1 | - | 10 | 1155 | |||
| JQ924062.1 | - | 10 | 1028 | |||

ATG16L1
Genomic Map