siRNA Details
| Gene Symbol: TEX13B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031273.2 | NP_112563.1 | 3 | 792 | ||
| GenBank mRNA | AF285598.1 | - | 3 | 792 | ||
| BC150140.1 | - | 1 | 1040 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEX13B| Gene Symbol: TEX13B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031273.2 | NP_112563.1 | 3 | 792 | ||
| GenBank mRNA | AF285598.1 | - | 3 | 792 | ||
| BC150140.1 | - | 1 | 1040 | |||