siRNA Details
| Gene Symbol: HIST1H4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003542.3 | NP_003533.1 | 1 | 177 | ||
| GenBank mRNA | BC054014.1 | - | 1 | 233 | 6499185 | |
| BC130558.1 | - | 1 | 249 | 40146589 | ||
| BC130560.1 | - | 1 | 249 | 40146591 | ||

HIST1H4C
Genomic Map