siRNA Details
| Gene Symbol: NXF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028089.1 | 19 | 1899 | |||
| GenBank mRNA | AJ277654.1 | - | 19 | 1899 | ||
| AJ277655.1 | - | 19 | 1874 | |||
| AJ277656.1 | - | 19 | 1810 | |||
| AJ277657.1 | - | 17 | 1685 | |||
| AJ277658.1 | - | 17 | 1804 | |||

NXF5
Genomic Map