siRNA Details
Gene Symbol: HIST1H4G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003547.2 | NP_003538.1 | 1 | 306 | ||
GenBank mRNA | BC126276.1 | - | 1 | 360 | 8991992 | |
BC126278.1 | - | 1 | 360 | 8991994 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HIST1H4G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003547.2 | NP_003538.1 | 1 | 306 | ||
GenBank mRNA | BC126276.1 | - | 1 | 360 | 8991992 | |
BC126278.1 | - | 1 | 360 | 8991994 |