siRNA Details
| Gene Symbol: MFN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033540.2 | NP_284941.2 | 18 | 2863 | ||
| XM_005247596.3 | XP_005247653.2 | 18 | 2821 | |||
| XM_011512963.2 | XP_011511265.1 | 15 | 2449 | |||
| GenBank mRNA | AK000700.1 | - | 17 | 2693 | ||
| BC040557.1 | - | 18 | 2863 | 5270347 | ||
| CR627379.1 | - | 15 | 2512 | |||

MFN1
Genomic Map