siRNA Details
| Gene Symbol: KLHL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019117.4 | NP_061990.2 | 2 | 661 | ||
| NM_057162.2 | NP_476503.1 | 2 | 661 | |||
| GenBank mRNA | AB051474.1 | - | 2 | 607 | ||
| AF284765.1 | - | 2 | 568 | |||
| AF284766.1 | - | 2 | 568 | |||
| AK022715.1 | - | 2 | 84 | |||
| AL832514.1 | - | 2 | 601 | |||
| BC140839.1 | - | 2 | 530 | 9021707 | ||
| BC140840.1 | - | 2 | 530 | 9021708 | ||
| BC142652.1 | - | 2 | 507 | 40148044 | ||
| BC146677.1 | - | 2 | 507 | 40148894 | ||
| BC161675.1 | - | 2 | 607 | 8862604 | ||

KLHL4
Genomic Map