siRNA Details
| Gene Symbol: SMIM11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017028203.1 | XP_016883692.1 | 5 | 602 | ||
| XM_017028204.1 | XP_016883693.1 | 4 | 413 | |||
| Gene Symbol: SMIM11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017028397.1 | XP_016883886.1 | 5 | 602 | ||
| XM_017028398.1 | XP_016883887.1 | 4 | 413 | |||
| GenBank mRNA | AK092693.1 | - | 5 | 578 | ||
| AY081143.1 | - | 4 | 426 | |||
| AY081144.1 | - | 5 | 593 | |||
| BC045820.1 | - | 4 | 414 | |||

SMIM11B
Genomic Map