siRNA Details
| Gene Symbol: LINC00158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024027.2 | 1 | 511 | |||
| GenBank mRNA | AY035382.1 | - | 1 | 510 | ||
| AY035383.1 | - | 1 | 510 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00158| Gene Symbol: LINC00158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024027.2 | 1 | 511 | |||
| GenBank mRNA | AY035382.1 | - | 1 | 510 | ||
| AY035383.1 | - | 1 | 510 | |||