siRNA Details
| Gene Symbol: EVA1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286556.1 | NP_001273485.1 | 1, 2 | 574 | ||
| NM_058187.4 | NP_478067.2 | 1, 2 | 574 | |||
| NR_135467.1 | 1, 2 | 630 | ||||
| XM_005261019.4 | XP_005261076.1 | 1, 2 | 695 | |||
| XM_006724038.3 | XP_006724101.1 | 1, 2 | 695 | |||
| XM_017028418.1 | XP_016883907.1 | 1, 2 | 695 | |||
| XM_017028419.1 | XP_016883908.1 | 1, 2 | 695 | |||
| GenBank mRNA | AF358258.1 | - | 1, 2 | 157 | ||
| AY358787.1 | - | 1, 2 | 567 | |||
| BC038710.1 | - | 1, 2 | 309 | 4813873 | ||

EVA1C
Genomic Map