siRNA Details
| Gene Symbol: TSPEAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272037.1 | NP_001258966.1 | 5 | 719 | ||
| NM_144991.2 | NP_659428.2 | 4 | 657 | |||
| GenBank mRNA | AJ487962.1 | - | 4 | 657 | ||
| BC021197.2 | - | 4 | 546 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSPEAR| Gene Symbol: TSPEAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272037.1 | NP_001258966.1 | 5 | 719 | ||
| NM_144991.2 | NP_659428.2 | 4 | 657 | |||
| GenBank mRNA | AJ487962.1 | - | 4 | 657 | ||
| BC021197.2 | - | 4 | 546 | |||