siRNA Details
| Gene Symbol: UBR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175748.3 | NP_786924.2 | 2 | 413 | ||
| GenBank mRNA | AK097477.1 | - | 2 | 398 | ||
| AK315492.1 | - | 2 | 212 | |||
| BC015046.1 | - | 2 | 212 | 3908032 | ||
| BC051819.1 | - | 2 | 175 | 6171393 | ||
| BX248249.1 | - | 2 | 172 | |||
| DB047433.1 | - | 2 | 413 | |||

UBR7
Genomic Map