siRNA Details
| Gene Symbol: UBR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175748.3 | NP_786924.2 | 8 | 1074 | ||
| NR_038150.1 | 7 | 940 | ||||
| GenBank mRNA | AK001345.1 | - | 5 | 452 | ||
| AK097477.1 | - | 8 | 1059 | |||
| AK315492.1 | - | 8 | 873 | |||
| BC015046.1 | - | 8 | 873 | 3908032 | ||
| BC017013.1 | - | 7 | 718 | 3883770 | ||
| BC051819.1 | - | 8 | 836 | 6171393 | ||
| BX248249.1 | - | 8 | 833 | |||

UBR7
Genomic Map