siRNA Details
Gene Symbol: PRKD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005813.4 | NP_005804.1 | 1 | 771 | ||
XM_005264237.3 | XP_005264294.1 | 2 | 1044 | |||
XM_017003741.1 | XP_016859230.1 | 2 | 994 | |||
GenBank mRNA | AB015982.2 | - | 1 | 771 | ||
BC012820.2 | - | 1 | 730 | |||
BC030706.2 | - | 1 | 716 | 5266769 |
