siRNA Details
| Gene Symbol: TRIM29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012101.3 | NP_036233.2 | 1 | 885 | ||
| XM_011542729.1 | XP_011541031.1 | 1 | 909 | |||
| XM_011542732.1 | XP_011541034.1 | 1 | 909 | |||
| GenBank mRNA | AF230388.1 | - | 1 | 885 | ||
| AF230389.1 | - | 1 | 885 | |||
| AK122720.1 | - | 2 | 701 | |||
| AK223090.1 | - | 1 | 843 | |||
| AK299472.1 | - | 1 | 242 | |||
| BC010644.1 | - | 1 | 831 | 3855016 | ||
| BC017352.2 | - | 1 | 824 | 5013158 | ||
| L24203.1 | - | 1 | 885 | |||

TRIM29
Genomic Map