siRNA Details
| Gene Symbol: FBXL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012158.2 | NP_036290.1 | 5 | 2430 | ||
| XM_005266336.1 | XP_005266393.1 | 5 | 2213 | |||
| XM_005266337.2 | XP_005266394.1 | 4 | 2065 | |||
| GenBank mRNA | AF126028.1 | - | 4 | 2035 | ||
| AK025484.1 | - | 3 | 1628 | |||
| AL833187.1 | - | 5 | 2378 | |||
| BC072448.1 | - | 5 | 2384 | 6166244 | ||

FBXL3
Genomic Map