siRNA Details
| Gene Symbol: FBXL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278716.1 | NP_001265645.1 | 4 | 618 | ||
| NM_012160.4 | NP_036292.2 | 3 | 500 | |||
| NR_103836.1 | 3 | 500 | ||||
| NR_103837.1 | 3 | 500 | ||||
| XM_005266930.2 | XP_005266987.1 | 4 | 606 | |||
| XM_011535748.2 | XP_011534050.1 | 4 | 606 | |||
| XM_017010726.1 | XP_016866215.1 | 3 | 432 | |||
| XM_017010727.1 | XP_016866216.1 | 3 | 460 | |||
| XM_017010728.1 | XP_016866217.1 | 4 | 618 | |||
| GenBank mRNA | AF174590.1 | - | 1 | 109 | ||
| AF176699.1 | - | 1 | 109 | |||
| AF199355.1 | - | 1 | 113 | |||
| AK313076.1 | - | 3 | 429 | |||
| BC055010.1 | - | 3 | 439 | 6137889 | ||
| BC091484.1 | - | 4 | 538 | 6178874 | ||
| BQ213738.1 | - | 3 | 479 | |||

FBXL4
Genomic Map