siRNA Details
| Gene Symbol: FBXO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297437.1 | NP_001284366.1 | 2 | 444 | ||
| NM_012176.2 | NP_036308.1 | 2 | 442 | |||
| NM_033484.2 | NP_277019.1 | 2 | 442 | |||
| XM_011514026.2 | XP_011512328.1 | 2 | 447 | |||
| XM_011514027.2 | XP_011512329.1 | 2 | 374 | |||
| GenBank mRNA | AF176703.1 | - | 2 | 380 | ||
| AK001003.1 | - | 1 | 1920 | |||
| AK024075.1 | - | 2 | 442 | |||
| AU143782.1 | - | 2 | 442 | |||
| BC040011.1 | - | 2 | 403 | |||
| BC048098.1 | - | 2 | 410 | 4830192 | ||

FBXO4
Genomic Map