siRNA Details
| Gene Symbol: FBXO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142522.1 | NP_001135994.1 | 4 | 1334 | ||
| NM_012177.3 | NP_036309.1 | 4 | 1174 | |||
| GenBank mRNA | AF129535.1 | - | 4 | 1126 | ||
| AJ420499.1 | - | 4 | 1194 | |||
| AK026197.1 | - | 5 | 1083 | |||
| AK055221.1 | - | 4 | 1308 | |||
| AK314456.1 | - | 4 | 1156 | |||
| AY079515.1 | - | 4 | 1065 | |||
| BC018905.1 | - | 4 | 1116 | 3960052 | ||

FBXO5
Genomic Map