siRNA Details
| Gene Symbol: FBXO7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033024.1 | NP_001028196.1 | 3 | 450 | ||
| NM_001257990.1 | NP_001244919.1 | 3 | 602 | |||
| NM_012179.3 | NP_036311.3 | 3 | 845 | |||
| XM_011530106.1 | XP_011528408.1 | 2 | 588 | |||
| GenBank mRNA | AF129537.1 | - | 2 | 459 | ||
| AF233225.1 | - | 3 | 565 | |||
| AK226087.1 | - | 3 | 760 | |||
| AK291972.1 | - | 3 | 585 | |||
| AK297841.1 | - | 3 | 405 | |||
| AK301716.1 | - | 3 | 530 | |||
| AL050254.1 | - | 3 | 767 | |||
| BC008361.1 | - | 3 | 842 | 3611049 | ||
| BI821903.1 | - | 3 | 450 | |||
| BX362053.2 | - | 3 | 817 | |||
| BX648151.1 | - | 3 | 398 | |||
| BX648220.1 | - | 3 | 395 | |||
| CR456491.1 | - | 3 | 680 | |||

FBXO7
Genomic Map