siRNA Details
| Gene Symbol: DNAJB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135004.2 | NP_001128476.2 | 5 | 1845 | ||
| NM_001135005.2 | NP_001128477.1 | 4 | 2052 | |||
| NM_012266.5 | NP_036398.3 | 4 | 1700 | |||
| XM_005251428.3 | XP_005251485.1 | 4 | 1619 | |||
| XM_006716751.2 | XP_006716814.1 | 5 | 1709 | |||
| XM_011517842.2 | XP_011516144.1 | 5 | 2286 | |||
| GenBank mRNA | AF087870.1 | - | 5 | 1702 | ||
| AK023253.1 | - | 4 | 1596 | |||

DNAJB5
Genomic Map