siRNA Details
| Gene Symbol: SLC39A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012319.3 | NP_036451.3 | 2 | 455 | ||
| XM_011525900.1 | XP_011524202.1 | 2 | 888 | |||
| XM_011525901.1 | XP_011524203.1 | 3 | 1572 | |||
| GenBank mRNA | AK304079.1 | - | 2 | 379 | ||
| DC410860.1 | - | 2 | 455 | |||
| DC414741.1 | - | 2 | 455 | |||
| U41060.2 | - | 3 | 302 | |||

SLC39A6
Genomic Map