siRNA Details
| Gene Symbol: IFIT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012420.2 | NP_036552.1 | 2 | 1193 | ||
| GenBank mRNA | AK293346.1 | - | 3 | 855 | ||
| AK312358.1 | - | 2 | 1026 | |||
| BC025786.1 | - | 2 | 1172 | 5229306 | ||
| CR457031.1 | - | 1 | 962 | |||
| U34605.1 | - | 2 | 1079 | |||

IFIT5
Genomic Map