siRNA Details
| Gene Symbol: ANKRD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256182.1 | NP_001243111.1 | 10 | 2550 | ||
| NM_001256183.1 | NP_001243112.1 | 9 | 2476 | |||
| NM_013275.5 | NP_037407.4 | 9 | 2479 | |||
| XM_006721181.1 | XP_006721244.1 | 9 | 2256 | |||
| XM_006721184.2 | XP_006721247.1 | 8 | 2294 | |||
| XM_011523051.2 | XP_011521353.1 | 10 | 2539 | |||
| XM_011523053.2 | XP_011521355.1 | 10 | 2645 | |||
| XM_011523054.2 | XP_011521356.1 | 10 | 2559 | |||
| XM_011523055.2 | XP_011521357.1 | 11 | 2844 | |||
| XM_011523056.2 | XP_011521358.1 | 10 | 2581 | |||
| XM_011523057.2 | XP_011521359.1 | 9 | 2462 | |||
| XM_017023182.1 | XP_016878671.1 | 9 | 2912 | |||
| XM_017023183.1 | XP_016878672.1 | 10 | 2648 | |||
| XM_017023184.1 | XP_016878673.1 | 10 | 2645 | |||
| XM_017023185.1 | XP_016878674.1 | 8 | 2286 | |||
| XM_017023186.1 | XP_016878675.1 | 12 | 3022 | |||
| XM_017023187.1 | XP_016878676.1 | 8 | 2374 | |||
| GenBank mRNA | AK290994.1 | - | 8 | 2158 | ||
| AY131227.1 | - | 1 | 875 | |||
| AY373756.1 | - | 9 | 2482 | |||
| AY533563.1 | - | 10 | 2307 | |||
| BC017437.1 | - | 9 | 2448 | |||

ANKRD11
Genomic Map