siRNA Details
| Gene Symbol: ZDHHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001323627.1 | NP_001310556.1 | 4 | 697 | ||
| NM_013304.2 | NP_037436.1 | 4 | 639 | |||
| XM_005255915.1 | XP_005255972.1 | 3 | 373 | |||
| XM_006721185.1 | XP_006721248.1 | 3 | 373 | |||
| XM_011523058.1 | XP_011521360.1 | 3 | 373 | |||
| XM_011523059.1 | XP_011521361.1 | 3 | 373 | |||
| XM_011523060.1 | XP_011521362.1 | 2 | 172 | |||
| GenBank mRNA | BC021908.1 | - | 4 | 665 | 4053374 | |
| U90653.2 | - | 4 | 639 | |||

ZDHHC1
Genomic Map