siRNA Details
| Gene Symbol: SNX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199835.1 | NP_001186764.1 | 7 | 1289 | ||
| NM_001199837.1 | NP_001186766.1 | 5 | 1094 | |||
| NM_001199838.1 | NP_001186767.1 | 4 | 993 | |||
| NM_001318198.1 | NP_001305127.1 | 8 | 1363 | |||
| NM_001318199.1 | NP_001305128.1 | 7 | 1164 | |||
| NM_013322.2 | NP_037454.2 | 7 | 1242 | |||
| XM_006715712.2 | XP_006715775.1 | 7 | 1442 | |||
| XM_017012085.1 | XP_016867574.1 | 9 | 1457 | |||
| XM_017012086.1 | XP_016867575.1 | 9 | 1896 | |||
| XM_017012087.1 | XP_016867576.1 | 8 | 1386 | |||
| XM_017012088.1 | XP_016867577.1 | 8 | 1333 | |||
| GenBank mRNA | AF121860.1 | - | 7 | 1155 | ||
| AK307905.1 | - | 5 | 1044 | |||
| AK316088.1 | - | 8 | 1339 | |||
| BC031050.1 | - | 7 | 1136 | 5278015 | ||
| BC034992.1 | - | 7 | 1215 | 4827599 | ||
| BX648363.1 | - | 1 | 2702 | |||

SNX10
Genomic Map