siRNA Details
Gene Symbol: GPR132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278694.1 | NP_001265623.1 | 5 | 1787 | ||
NM_001278695.1 | NP_001265624.1 | 3 | 917 | |||
NM_001278696.1 | NP_001265625.1 | 2 | 803 | |||
NM_013345.3 | NP_037477.1 | 4 | 1697 | |||
GenBank mRNA | AB465599.1 | - | 4 | 1577 | ||
AB465600.1 | - | 3 | 797 | |||
AF083955.1 | - | 4 | 1565 | |||
AK292142.1 | - | 5 | 1660 | |||
AK303654.1 | - | 3 | 797 | |||
AX775841.1 | - | 3 | 812 | |||
AX775935.1 | - | 3 | 797 | |||
AX775939.1 | - | 3 | 1275 | |||
BC084546.1 | - | 2 | 1252 | 5455247 | ||
BT007257.1 | - | 1 | 101 | |||
EU432121.1 | - | 2 | 665 | |||
EU543705.1 | - | 4 | 786 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
