siRNA Details
| Gene Symbol: TMOD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013353.2 | NP_037485.2 | 3 | 337 | ||
| XM_011509449.1 | XP_011507751.1 | 3 | 219 | |||
| XM_017001089.1 | XP_016856578.1 | 2 | 954 | |||
| XM_017001090.1 | XP_016856579.1 | 3 | 266 | |||
| GenBank mRNA | AF165217.1 | - | 3 | 214 | ||
| AF177173.2 | - | 3 | 282 | |||
| AK300675.1 | - | 3 | 247 | |||
| AK313280.1 | - | 3 | 246 | |||
| BC017810.1 | - | 3 | 247 | 4288769 | ||
| BX647346.1 | - | 3 | 247 | |||
| DC385872.1 | - | 3 | 337 | |||

TMOD4
Genomic Map