siRNA Details
| Gene Symbol: CYTH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318024.1 | NP_001304953.1 | 5 | 506 | ||
| NM_013385.4 | NP_037517.1 | 5 | 487 | |||
| XM_011530147.2 | XP_011528449.1 | 5 | 374 | |||
| XM_011530148.1 | XP_011528450.1 | 5 | 336 | |||
| GenBank mRNA | AF075458.1 | - | 5 | 416 | ||
| AF125349.1 | - | 5 | 487 | |||
| AK024428.1 | - | 5 | 341 | |||
| AK304453.1 | - | 5 | 407 | |||
| AK315052.1 | - | 5 | 323 | |||
| BC017780.1 | - | 5 | 355 | |||
| BC041161.1 | - | 5 | 345 | 5745783 | ||
| CR456551.1 | - | 5 | 336 | |||

CYTH4
Genomic Map