siRNA Details
| Gene Symbol: CYTH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318024.1 | NP_001304953.1 | 13 | 2296 | ||
| NM_013385.4 | NP_037517.1 | 13 | 2277 | |||
| XM_011530147.2 | XP_011528449.1 | 13 | 2164 | |||
| XM_011530148.1 | XP_011528450.1 | 14 | 2241 | |||
| GenBank mRNA | AF075458.1 | - | 13 | 2206 | ||
| AK002145.1 | - | 7 | 1593 | |||
| AK024428.1 | - | 12 | 3536 | |||
| BC041161.1 | - | 13 | 2135 | 5745783 | ||

CYTH4
Genomic Map