siRNA Details
Gene Symbol: ADGRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271052.1 | NP_001257981.1 | 18, 19 | 2467 | ||
NM_013447.3 | NP_038475.2 | 20, 21 | 2926 | |||
GenBank mRNA | AF114491.1 | - | 19, 20 | 2539 | ||
AK024426.1 | - | 10, 11 | 1295 | |||
AK024454.1 | - | 5, 6 | 508 | |||
AK291776.1 | - | 19, 20 | 2672 | |||
AK298700.1 | - | 18, 19 | 2383 | |||
BC034949.1 | - | 15, 16 | 2673 | |||
BC053933.1 | - | 15, 16 | 2666 | |||
BC082991.1 | - | 19, 20 | 2483 | |||
BC127004.1 | - | 20, 21 | 2922 | 40108332 | ||
BC127005.1 | - | 20, 21 | 2922 | 40108333 | ||
CR936819.1 | - | 14, 15 | 1903 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
