siRNA Details
Gene Symbol: ADGRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271052.1 | NP_001257981.1 | 19 | 2843 | ||
NM_013447.3 | NP_038475.2 | 21 | 3302 | |||
GenBank mRNA | AF114491.1 | - | 20 | 2915 | ||
AK024426.1 | - | 11 | 1671 | |||
AK024454.1 | - | 6 | 883 | |||
BC034949.1 | - | 16 | 3049 | |||
BC053933.1 | - | 16 | 3042 | |||
BC082991.1 | - | 20 | 2859 | |||
CR936819.1 | - | 15 | 2279 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
