siRNA Details
| Gene Symbol: CLIC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013943.2 | NP_039234.1 | 4, 5 | 698 | ||
| GenBank mRNA | AF097330.1 | - | 4, 5 | 553 | ||
| AF109196.1 | - | 4, 5 | 491 | |||
| AK001624.1 | - | 4, 5 | 585 | |||
| AK095959.1 | - | 4, 5 | 519 | |||
| AK301466.1 | - | 4, 5 | 496 | |||
| AK307142.1 | - | 2, 3 | 263 | |||
| AK315051.1 | - | 4, 5 | 535 | |||
| AL117424.1 | - | 4, 5 | 610 | |||
| BC012444.1 | - | 4, 5 | 563 | 3861372 | ||
| CR533501.1 | - | 4, 5 | 413 | |||

CLIC4
Genomic Map