siRNA Details
| Gene Symbol: OSTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014028.3 | NP_054747.2 | 3 | 648 | ||
| GenBank mRNA | AF533891.1 | - | 3 | 632 | ||
| AK075012.1 | - | 3 | 574 | |||
| AK129854.1 | - | 3 | 642 | |||
| AK314393.1 | - | 3 | 324 | |||
| AY358795.1 | - | 3 | 639 | |||
| BC068581.1 | - | 3 | 630 | 5298450 | ||
| BI668275.1 | - | 3 | 665 | |||

OSTM1
Genomic Map