siRNA Details
| Gene Symbol: SNX24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014035.2 | NP_054754.1 | 7 | 897 | ||
| XM_017009394.1 | XP_016864883.1 | 7 | 722 | |||
| GenBank mRNA | AF139461.1 | - | 7 | 786 | ||
| AK021473.1 | - | 1 | 1363 | |||
| BC010886.1 | - | 7 | 857 | 3538426 | ||
| BC069012.1 | - | 7 | 920 | 4585781 | ||

SNX24
Genomic Map