siRNA Details
| Gene Symbol: CD274 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267706.1 | NP_001254635.1 | 6 | 754 | ||
| NM_014143.3 | NP_054862.1 | 7 | 1096 | |||
| NR_052005.1 | 8 | 923 | ||||
| GenBank mRNA | AF233516.1 | - | 7 | 1040 | ||
| AK001894.1 | - | 5 | 803 | |||

CD274
Genomic Map