siRNA Details
| Gene Symbol: MKRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271707.1 | NP_001258636.1 | 6 | 1038 | ||
| NM_014160.4 | NP_054879.3 | 7 | 1167 | |||
| GenBank mRNA | AF302084.1 | - | 7 | 1081 | ||
| AK091318.1 | - | 7 | 1112 | |||
| AK298463.1 | - | 6 | 965 | |||
| AL834512.1 | - | 4 | 689 | |||
| BC001799.2 | - | 5 | 876 | |||
| BC015715.1 | - | 7 | 1109 | 3916199 | ||

MKRN2
Genomic Map